Research Application
|
The research application in use in the analysis workflow.
|
Workflow name
|
The name and version of the analysis workflow.
|
Description
|
A short description of the analysis workflow.
|
Version
|
The version number of Ion Reporter™ Software.
|
Revision
|
The revision number of the analysis workflow.
|
Sample Group
|
The sample group in use in the analysis workflow.
|
Research Category
|
The research category selected for the analysis workflow.
|
Reference
|
The reference selected for the analysis workflow.
|
Target Regions
|
The filename of the target regions file used in the analysis workflow.
|
Variant Type Detection
|
The variant type detection selected for the analysis workflow.
|
Hotspot Regions
|
The filename of the hotspots file used in the analysis workflow.
|
Fusion Panel
|
The name and version of the fusion panel.
|
MSI Baseline File
|
The filename of the MSI baseline file used in the analysis workflow. This file contains baseline information to account for variations in sample preparation or run conditions to ensure robust MSI detection.
|
MSI Marker Regions
|
The filename of the MSI marker regions file used in the analysis workflow. The MSI marker regions file contains information about the genomic position of the MSI markers, and additional information that is used by the MSI detection algorithm.
|
TMB Filter Chain
|
The TMB Filter Chain that is used in the analysis workflow.
|
TMB Calculation Version
|
The version of the tumor mutational burden algorithm that is available in the analysis workflow is listed. The algorithm version can provide information about how the data was analyzed and can be useful if earlier versions of the software were used, or in the case of troubleshooting.
|
Sequence Variant Baseline
|
The filename of the Sequence Variant Baseline file used in the analysis workflow.
|
Annotations
|
The annotations that are used in the analysis workflow.
|
MyVariants Database
|
The MyVariants database in use in the analysis workflow.
|
Filter Chain
|
The filter chain that is in use for the analysis workflow.
|
Copy Number
|
The copy number baseline control in use in the analysis workflow. The copy number baseline identifies the copy number variants (CNVs) in samples.
|
Plugins
|
The name and version number of the plugin or plugins in use in the analysis workflow.
|
Report Template
|
The name of the report template used in the analysis workflow.
|
Modified By
|
The Ion Reporter™ Software user who modified the analysis workflow.
|
Modified On
|
The date and time that the analysis workflow was modified.
|
Created By
|
The Ion Reporter™ Software user who created the analysis workflow.
|
Created On
|
The date and time that the analysis workflow was created.
|
Parameters
|
Click the View link to open a window to see all of the parameters that are defined for the selected analysis workflow.
|
Tag for IRU
|
Indicates whether the analysis workflow is tagged for use with IonReporterUploader plugin.
|