This filter can be added to filter chains in the software to detect variants that
match those in the Online Mendelian Inheritance in Man® (OMIM) database. The specific
filters are:
OMIM(20180823)
OMIM(20191001)
Filter conditions include the ability to search for all (In), none (Not In), or
specific annotations, and to include or exclude unannotated variants in filtered
analysis results.
The filter returns variants in samples that match those in the OMIM database.