Example Coverage Imbalance Scores plot
RNA Exon Tile Fusion Imbalance plots provide a visual representation of the RNA fusion imbalance analysis results for the detection of novel fusions in a partner agnostic manner. The Coverage Imbalance Scores plot shows a heatmap of imbalance scores for all selected analyses. Each row corresponds to a specific sample and each column corresponds to a specific gene. The Coverage Imbalance Scores plot is generated for multi-analysis comparison only, and does not take into account the p-values. To determine the significance of the imbalance score, review the Fusion Imbalance Call: Imbalance Score & P-value plot and Sample Coverage for Imbalance Analysis plot for each individual sample. For more information, see Example RNA Exon Tile Fusion Imbalance plots for a single analysis and Example sample coverage plot for a single analysis positive for a fusion imbalance in NTRK1.