Transfer multiple samples to a user in another organization
To share multiple samples with a user in another Ion Reporter™ Software organization, you can transfer a batch of samples to the user.
  1. In the Samples tab, click Overview.
  2. Select multiple samples, then click ActionsTransfer.
  3. In the Batch Transfer dialog box, enter the email address of the intended recipient, then click Transfer.
    A status appears stating your samples transfer has completed.
The recipient receives a notification and can download the transferred samples from the Notifications page. To free up storage space, you can delete the samples after transfer. For more information, see Delete a sample.