Run IonReporterUploader command-line utility
The IonReporterUploader command-line utility irucli is ready to run after you extract it. Run the IonReporterUploader command-line utility from the IonReporterUploader-cli bin directory (with the irucli.bat or irucli.sh script).
Instructions to use the IonReporterUploader command-line utility on a standard Linux™ computer are downloaded with the utility.
For more information, see the IonReporterUploader Command-Line Utility User Guide (Pub. No MAN0017648.)