IonReporterUploader command-line utility
You can use the IonReporterUploader command-line utility to transfer or upload sequencing run results files to Ion Reporter Software, then define samples that are based on the transferred files. The IonReporterUploader command-line utility uses your Ion Reporter Software account credentials to transfer run data from Torrent Suite Software to the organization on the specified Ion Reporter Server. The utility, which is available at http://iru.ionreporter.thermofisher.com/, supports these types of file transfers:
  • Transfers of a single BAM or VCF file.
    The name of a BAM file that is uploaded to Ion Reporter Software cannot exceed 200 characters.
  • Transfers of all results files for a Torrent Suite Software analysis.
  • Transfers of results files that are in a single flat folder.
  • Transfers of multiple files that are not restricted to a single folder.
  • Uploads of samples from hg19 and GRCh38 reference genomes only. Although the E-coli and animal reference genomes can be used in Torrent Suite Software, these reference genomes are not supported in Ion Reporter Software.
  • To upload samples from other reference genomes, you can build a plugin that includes any reference genome. For more information, see Programming guidelines for Ion Reporter Software plugins.
You can later analyze the samples with Ion Reporter Software. The IonReporterUploader command-line utility can be run on your Torrent Server, or can be run on any standard (and current) computer that runs Linux, macOS, or Windows Operating System. You can enter IonReporterUploader command-line utility options through command-line arguments, or through a properties file.
The IonReporterUploader command-line utility supports the upload of combined run results that are output by using the Combine Alignments option in the Torrent Suite Software. Uploads of combined run results are not supported when the IonReporterUploader plugin is used in Torrent Suite Software.
For more information on the IonReporterUploader command-line utility, see the IonReporterUploader Command-Line Utility User Guide (Pub. No. MAN0017648).