Example RNA Exon Tile Fusion Imbalance call plot for multiple analyses
RNA Exon Tile Fusion Imbalance plots provide a visual representation of the RNA fusion imbalance analysis results for the detection of novel fusions in a partner agnostic manner. The Multi Analysis Fusion Imbalance Calls: Imbalance Score & P-value plot shows the imbalance scores and p‑values for all the genes in all the selected analyses. Each data point corresponds to a gene-sample pair. Points that have the same color represent the same gene and are defined in the legend on the right. Points that have the same shape represent genes from the same sample and are defined in the legend on the bottom. This plot provides an overview of the relative confidence and strength of the fusion imbalance call only. To confirm the fusion imbalance call for any given gene, review and verify the imbalance score and P value for any given gene in the visualizations for a single analysis. For more information, see Example RNA Exon Tile Fusion Imbalance plots for a single analysis and Example sample coverage plot for a single analysis positive for a fusion imbalance in NTRK1.