You can review an existing custom and predesigned report templates.
In the Workflows tab, click Presets.
In the first dropdown list, select Final Report Templates, then, select a template type.
Option
Description
System-installed report templates
System-installed report templates are labeled and cannot be
altered. However, you can review the components of the factory
templates in the
Details
pane.
Custom templates
Custom templates and can be added to analysis workflows, deleted, edited, or locked.