Set password policies
An administrator is responsible for setting password policies for all Ion Reporter™ Software users. The software allows for strong password authentication (combination of numbers and letters and special characters), and requires password rotation every 90 days.
This feature is available in Ion Reporter™ Server and not available in Ion Reporter™ Software on Thermo Fisher™ Connect Platform.
This feature allows you to meet Title 21 CFR Part 11 of Federal Regulations that establishes the United States Food and Drug Administration regulations on electronic records and signatures, password policies, and user activity auditing.
- In the Admin tab, click Policies.
- In the Password Policy section, specify the password policy values and selections for all Ion Reporter™ Software users.
- Click Save.
×