Detailed analysis metrics
Metric
Description
Sample ID
Name of the sequenced sample imported from a sequencing run.
SNV/INDEL
Gene
HGNC reviewed official gene symbol.
AA Chg
Amino acid change resulting from non-synonymous DNA variant.
Mutant Frequency %
Frequency of mutant allele expressed as a percentage.
Oncomine Variant Class
Variant class annotation that provides a high-level summary of the variant type (hotspot, fusion, amplification, and so on). The annotation is included when the Oncomine™ Variant Annotator plugin is run in Ion Reporter™ Software.
Oncomine Gene Class
Variant gene functional annotation that provides a high-level summary of gene type (Gain-of-Function, Loss-of-Function, or Unclassified). The annotation is included when the Oncomine™ Variant Annotator plugin is run in Ion Reporter™ Software.
Info
HS (targeted hotspot) or PN (potentially novel TP53 variant). De novo variant calls available for the breast panel only.
Genotype
Genotype measured associated with a DNA variant call.
Ref Allele
Reference allele as defined in the human genome reference (hg19).
Mut Molecular Cov.
Molecular coverage of the mutant allele.
WT Molecular Cov.
Molecular coverage of the wild type allele from the reference genome.
Depth
Total read coverage across amplicon containing SNV/INDEL hotspot locations. Count of chip-level reads aligned at this locus that participate in variant calling.
QC Test (LOD) %
Quality control check for SNV/INDEL target regions based on molecular coverage.
Transcript ID
NCBI accession number for the transcript representing the gene target being measured.
Locus
Chromosome and position of detected variant. Click the hyperlink to open the Ion Reporter™ Genomic Viewer to the specified locus.
CNV
Gene
Gene locus targeted for CNV measurement.
Gain/Loss
Detected copy number gain or loss.
CNV ratio
Ratio of measured CNV gene locus coverage relative to coverage on non-CNV loci.
p-value
Significance of CNV ratio measurement.
Med. Mol Cov. Gene
Median molecular coverage of targeted CNV gene.
Med. Mol Cov. Ref
Median molecular coverage of non-CNV reference loci.
Med. Read Cov. Gene
Median read coverage of targeted CNV gene.
Med. Read Cov. Ref
Median read coverage of non-CNV reference loci.
QC Test
Assay quality control as determined by amplicon coverage uniformity and number of amplicons remaining after outlier removal.
Valid CNV Amplicons
Number of CNV amplicons remaining after outlier removal.
CNV Locus
Chromosomal location of CNV gene being targeted.
Fusion
Variant (exons)
Name of fusion targeted and respective acceptor and donor exons.
Oncomine Driver Gene
The gene thought to be associated with increased oncogenic properties due to inappropriate activation by the fusion.
COSMIC/NCBI
COSMIC mutation or NCBI accession number.
Mol Cov. Mutant
Median molecular coverage across fusion amplicon.
Read Cov. Mutant
Median read coverage across fusion amplicon.
Detection
Detection status from assay.
QC Test
Assay quality control measured from expression detection of housekeeping genes.
Type
Assay type (For example, Fusion, RNA exon variant (exon skipping), Proc Control).
Locus
Chromosomal locations of targets included in assay.
Ratio To Wild Type
Ratio molecular for exon skipping assay relative to wild type control amplicons.
Norm count Within Gene
Exon skipping assay coverage normalized to molecular coverage of wild type (WT) MET control amplicons. (Lung panel only).