Use filter chains to change analysis results
You can review the results of an Oncomine™ analysis run through a TagSeq analysis workflow, then apply different filter chains to alter the results.
-
In the Analyses tab, click the link in the Analysis column to open an analysis of interest.
The analysis results appear with the Oncomine™ Variants filter chain applied.
- (Optional) In the Filter Options pane, select a different filter chain from the list.
- Review the results.
×