Reset the API token for Ion Reporter™ Software on Connect
These procedure can be performed by either an administrative user or a regular user. To reset the API token for Ion Reporter™ Software on Connect, see instead Reset the API token on Ion Reporter Server.
The API token for Ion Reporter™ Software on Connect is used for:
  • Configuration of the IonReporterUploader plugin in in Torrent Suite™ Software.
  • The access of data from the Web services API.
Two functions of the IonReporterUploader plugin do not work when you regenerate the API token. The functions and the steps to recover functionality are as follows:
  • IonReporterUploader plugins that are configured with your Ion Reporter™ Software on Connect account.
  • Scripts or code that access the Ion Reporter™ Software on Connect Web services API with your previous token. To recover, use your new token with those scripts.
You must have an access code to complete this procedure if you use Ion Reporter™ Software on Connect.
If you use Ion Reporter™ Software on Connect and do not already have an access code, regenerate an access code and access the API token. The access code must contain at least 6 characters. The maximum length of the access code is 50 characters.
  1. Sign into Ion Reporter™ Software on Connect.
  2. Click (Settings) Manage Tokens.
  3. Click Set New Access Code, then enter an access code in the New Access code field.
  4. Select an expiration time in the Access code Age dropdown menu, then click Save and Generate.
  5. Click Copy to copy the API token to the clipboard.
The IRU token is for use with the IonReporterUploader command-line utility and is not required for this procedure.
A new API token is generated.
  • Use the new API token to reactivate the account that is configured for use with the IonReporterUploader plugin in Torrent Suite™ Software.
    In Torrent Suite™ Software when you set up an account for use with the IonReporterUploader plugin, you must enter the password in the Password field, then click Update.
    When you set up an account for use with the IonReporterUploader plugin, enter the access code in the Password field, then click Update.
  • Use the new API token with scripts or code that access the Ion Reporter™ Software on Connect Web services API.