Download an audit log for administrative policies
An administrator can download an audit log that records changes that are made to administrative policies in Ion Reporter™ Software.
This feature allows you to meet Title 21 CFR Part 11 of Federal Regulations that establishes the United States Food and Drug Administration regulations on electronic records and signatures, password policies, and user activity auditing.
- In the Admin tab, click Users.
- Click Download Audit Trail.
- Complete the Download Audit Trail dialog box.
- Click Download.
The audit log is downloaded through the browser. Click to save or open the file, depending on your browser settings.