Download an audit trail of user activities
An administrator can track the activities of individual users within the organization in Ion Reporter Software. Using this feature, you can select a screen and a time period to audit, then review a log file of the activities performed by users during that time period. For example, you can see who created analyses and who modified samples.
This feature allows you to meet Title 21 CFR Part 11 of Federal Regulations that establishes the United States Food and Drug Administration regulations on electronic records and signatures, password policies, and user activity auditing.
  1. In the Admin tab, click Users.
  2. Click Download Audit Trail.
  3. Complete the Download Audit Trail dialog box.
    1. In Screen Name, select a screen of interest from the list.
    2. In Date Range, select a date range, or specify from and to dates using the calendar.
    3. Click Download.
      A CSV file is created.
  4. Open the CSV file, then review user activities performed on the selected screen and date.
    Column
    Description
    Name
    Name of the analysis, analysis workflow, sample, or other item created or edited by the selected user.
    Type
    Type of item created or edited, such as analysis, MyVariants, and so on.
    Created By
    First and last name of the user who created an event.
    Created By Email
    Email address of the user who created an event.
    Updated By
    First and last name of the user who updated an event.
    Updated By Email
    Email address of the user who updated an event.
    Updated By Organization
    Name of the organization that updated an analysis or other element.
    Screen
    The name of the screen that was modified.
    Values
    The values that were changed on the modified screen.