Analyses tab overview
You can access the list of available analyses in the Analyses table, located on the in the Overview screen in the Analyses tab.
The Overview screen in the Analyses tab. Click the screen name to toggle between screens.
The Launch screen. For more information, see Launch an analysis.
The MyVariants screen. For more information, see MyVariants.
You can enter keywords or text strings in the Search box. For more information, see Search for analyses.
The Version All filter allows you to search for specific versions of analyses.
The Workflow All filter allows you to search for specific analysis workflows.
More Filters allows you to add additional filters to the Analyses tab. For more information, see Filter analyses.
Refresh refreshes the list of samples.
Apply Preferences to customize how the Analyses table is organized. For more information, see Customize the Analyses table.
Visualize allows you to view the data in Ion Reporter™ Genomic Viewer (IRGV) or Integrative Genomics Viewer (IGV). For more information, see Visualize analysis results with Ion Reporter Software.
Launch Analyses allows you to manually launch one or more analyses (batch). For more information, see Launch an analysis.
Additional filters, added by using More Filters.
Table columns with information about the analysis and data for analysis attributes. Several columns are included by default, and you can add more attributes with table preferences. For more information, see Analyses table column options.
(Checked Out) Indicates that edits to the analysis are in progress. Open the analysis in view-only mode to view analysis results and see the name of the user who is making edits to the analysis.
(Analyze role) Indicates that the analysis is shared by an Analyze role user.
(Notes) Notes about the analysis.
Details provides additional information about the selected analysis.