Add an analysis note
One or many analysis notes can be added to an analysis in Ion Reporter™ Software. An analysis note is any written comment or observation that you want to add to an analysis that has successfully completed. Anyone who is assigned Admin, Analyze, Import, or Report roles can add analysis notes.
You cannot edit analysis notes. However, you can add additional analysis notes to an analysis. You can also add a description to an analysis. For more information, see Launch an analysis.
Analyses notes can be shared within the same organization. Analysis notes are not shared when an analysis is shared with another Ion Reporter™ Software organization.
- In the Analyses tab, select Overview.
-
In the row of an analysis of interest, click
(Add) to add a new note, or
(Note) to add a note to the list of existing notes.
- In the Analysis Notes dialog box, click the Notes tab and enter a note in the text field.
- Click Add, then click Done.
The analysis note appears in the Details pane of a selected analysis on the Analyses Overview screen.