Search for an analysis note
You can search for the text in a specific analysis note or notes in Ion Reporter™ Software.
Analysis notes record any written comments or observations for an analysis that has successfully completed.
In the Analyses tab, click Overview.
|
Option |
Description |
|---|---|
|
To find one or more analysis notes that contain a specific word or phrase |
In the Search field, enter a word or phrase that is used in the analysis note, then click Go. |
|
To find all analysis notes |
(Optional) In the Search field, enter Note, then click Go. |