Ion AmpliSeq and Ion AmpliSeq HD panel types
AmpliSeq.com offers the following design and ordering options for Ion AmpliSeq and Ion AmpliSeq HD research panels.
  • Ion AmpliSeq On Demand panels of optimized amplicons for germline analysis. These panels are configurable to a specific human disease area, and ordered in small reaction packs.
  • Made-to-Order Ion AmpliSeq Custom DNA panels for germline or somatic analysis of any genome, and Made-to-Order Ion AmpliSeq Custom RNA panels for analysis of human gene expression and gene fusion events. These panels are ordered in large reaction packs.
  • Made-to-Order Ion AmpliSeq HD Custom ultra high-sensitivity DNA and RNA panels for liquid biopsy or somatic analysis of human DNA variants and RNA fusion events, including UMTs (Unique Molecular Tags) and dual barcodes.
  • Oncomine tumor specific panels, optimized for use with FFPE-derived DNA and RNA. Cancer-type specific core panels can be customized by adding or removing gene targets from inventory. Each panel consists of 2 DNA primer pools and an optional RNA pool for fusion event detection.
  • Ion AmpliSeq Kitted Panels, predesigned DNA and RNA panels for different applications. These panels are ordered in small reaction packs.