Audit logs
Audit logs record user activities, such as who launches analyses, the start and end time of analyses, report creation and publication, and edits of analyses, including the addition of annotations, MyVariants flags, classifications, and notes.
This feature allows you to meet Title 21 CFR Part 11 of Federal Regulations that establishes the United States Food and Drug Administration regulations on electronic records and signatures, password policies, and user activity auditing.