Available report sections
The following sections are available to add to report templates, final reports, or visualization reports.
More sections might be available in a visualization report. For more information, see Create a visualization report.
|
Section |
Description |
|---|---|
|
Organizational Information |
|
|
Background |
Enter background information. |
|
Analysis Information |
Generates a list of details for the analysis.
|
|
Samples Overview |
Includes all sample attributes that are included in the Samples table when the data are viewed in Ion Reporter™ Software. |
|
QC Metrics |
Generates a table coverage metrics for hotspots, amplicons and genes. For a complete list of the metrics that are included, see Quality control (QC) metrics. You can also view and download these metrics in a separate report. For more information, see Create a final report template from analysis results |
|
Reported Variants |
Generates a table of reported variants and includes columns of variant details. |
|
Variant Details |
Includes name and description of variants, and which samples they came from. In addition, it lists Annotations and Notes for each variant. |
|
Molecular Coverage Analysis |
Metrics that describe the level of sequence coverage for target genomic regions. The Coverage Metrics section is available if the coverageAnalysis plugin was run in Torrent Suite™ Software, before data was transferred to and analyzed in Ion Reporter™ Software. See Molecular Coverage Analysis metrics for more information. |
|
Coverage Analysis Metrics |
Metrics for molecular coverage over the targeted regions of the reference genome. The Molecular Coverage Analysis section is available if the molecular Coverage Analysis plugin was run in Torrent Suite™ Software, before data was transferred to and analyzed in Ion Reporter™ Software. See Coverage analysis metrics for more information. |
|
Comments |
Enter comments about the report or report contents. |
|
Sign–Off |
This section provides lines with the name and qualifications of people who are required to provide a handwritten signature for the report. Enter the name and title for a handwritten signature, then click Add. Continue to add the information for all required signatures, until all designated signers are added. |
|
Disclaimer |
Enter a custom legal disclaimer. |
|
Image |
You can add an Image section to a final report or final report template. If included, the Image section can be used to upload one or more images in PNG, GIF, JPG, or JPEG formats into the report. There is no limit to the number of Image sections that can be added. |
|
Footer |
You can remove the Thermo Fisher Scientific logo from the report footer and add a custom logo. Acceptable formats are PNG, GIF, and JPG. |