Sample presets
You can create user-defined sample attributes, which are known as sample presets, to describe samples that are used in research.
Ion Reporter™ Software supports the following types of attributes:
  • Boolean—Creates a list of these two choices: True, False.
  • Text—Accepted values: general text.
  • Integer—Accepted values: integer numbers. Examples: 0, 9, −44.
  • Float—Accepted values: decimal numbers Examples: 0.1, 9.0, −44.3.
  • Controlled Vocabulary—Creates a list of these choices that you define with the preset terms.
  • Date—Accepted values: provides a calendar tool to select a date.
Ion Reporter™ Software gives you the option to encrypt the internal storage for a sample attribute.