Transfer samples to users in other organizations
You can share samples with other users in different Ion Reporter™ Software organizations. To share a sample, you can transfer either a single sample or a batch of samples. Any user-defined attributes that are associated with the samples are also transferred with each sample. After a sample is transferred, you can delete the sample to free up storage space.