Accept a sample from a user in another organization
You can accept samples that are transferred from a user in another Ion Reporter™ Software organization.
The Ion Reporter™ Software account that you use for this procedure must include the Import, Analyze, or Admin roles.
  1. In the Home tab, click Notifications.
  2. Find the transferred sample, then click  (Download).
  3. Click the Samples tab, then find the transferred sample.
  4. To accept the sample, in the Details pane, click ActionsAccept.
The sample is available for use in the Samples table.