Transfer a sample to users in another organization
You can transfer samples to other users in different Ion Reporter™ Software organizations. All custom sample attributes are also transferred with the sample.
In the Sample tab, click Overview.
Click the row of the sample that you want to transfer, then click Actions > Transfer.
In the Transfer Sample dialog box, enter the address of one or more intended recipients, then click Transfer.
A transfer initiation message appears.
The recipient receives a notification and can download the transferred sample from the Notifications page.
To free up storage space, you can delete the sample after the transfer. For more information, see Delete a sample.
IMPORTANT! To avoid data loss, ensure that the user with whom you share the samples has accepted the transfer of the samples before you delete any samples.