Archive samples to an external storage device
This information applies only to Ion Reporter™ Server.
You can archive samples from an Ion Reporter™ Server to an external storage device to increase disk space.
Available options for external storage devices include the following.
-
Torrent Storage™ NAS (Network Attached Server) device (Network Attached Server)—For more information, see https://www.thermofisher.com/order/catalog/product/A32198
-
DataSafe™ Solution—For more information, see https://www.thermofisher.com/order/catalog/product/A32633
-
You need to have an external storage device that is mounted to your Ion Reporter™ Server to archive samples to the device.
-
If you have a new Ion Reporter™ Server or upgrade from a previous version, you need to specify a storage location for archiving samples and analyses in Ion Reporter™ Server.
IMPORTANT! Change archivalMountPath=/tmp/ to archivalMountPath=/storage IP address/ in the ionreportermanager/server/server.properties file, then restart the Tomcat server. To check the data that you archived, go to the storage location of the archived data, which is set up in /share/apps/IR/ionreportermanager/server/server.properties file as archivalMountPath. If you have questions about the Tomcat server, contact your Field Support Engineer or Field Bioinformatics Support representative.
- You can only archive samples if no processes, such as an analysis is running, or new sample uploads in progress, are active on the server.
- In the Samples tab, in the Overview screen, select the sample or samples to be archived.
-
Complete one of the following actions.
Option
Action
Archive multiple samples
In the Selected Samples section, click . Archive a single sample
In the Details section, click . -
Indicate whether the archives should be retained on the server or deleted in the Confirm Batch Archive dialog box.
- To delete the samples after the archival process is complete, select the option do you want to delete the data from path post archive?.
- To retain the samples after the archival process is complete, deselect the option do you want to delete the data from path post archive?.
-
Click Yes in the Confirm Batch Archive dialog box to begin the archive process for the selected samples.
The Archival Manager screen shows the progress of the archive.
-
(Optional) To stop the archive process, click Abort.
To prevent a loss of data, if you archive multiple analysis you can only abort the archive process if none of the archival processes are complete. For example, if there are 3 archives that are in progress, and 2 archives are completed, but the third item is in progress, the abort does not occur.