Add attributes to the Samples table
You can add sample attributes to the Samples table that you view in Ion Reporter™ Software.
  1. In the Samples tab, click Overview.
  2. Click PreferencesSelect Columns, then select the sample attributes that you want to add to the Samples table, or deselect the sample attributes that you want to remove from the table.
    User-defined attributes are denoted with an asterisk (*) in the available columns list.
The selected sample attributes are added to the Samples table until you close the table. If you create a sample CSV file before you close the table, each user-defined attribute is included in the CSV file.
To save the table with the attributes that you added, see Create a Samples table preference.