Predefined filters Ion Reporter™ Software includes predefined filters that you can apply to analysis workflows, analysis results or visualizations. The filters are categorized as annotation source filters and data type filters.