Data type filters
You can create filter chains that identify variants based on variant data types in the software, such as allele frequency and allele ratio.
Filter chains that include data type filters narrow the list of variants that appear in the analysis results, when applied to an analysis workflow, or when used to change the variants shown when you view or visualize analysis results.