CNV Confidence Range—CNVs Only filter
This filter can be added to filter chains in the software to return copy number variants (CNV) with confidence levels between 10 and 10,000,000.
Filter conditions include the ability to set the CNV confidence range from 10 to 1.0E7. In addition, the range boundary values can be included or excluded, and unannotated variants can be included or excluded in filtered analysis results.
The filter returns only copy number variants with high confidence levels.