Variant Type filter
This filter can be added to filter chains in the software to detect variants that match the following variant types:
-
SNV
-
INDEL
-
MNV
-
REF
-
NOCALL
-
CNV
-
LONGDEL
-
FUSION
-
EXPR_CONTROL
-
ASSAYS_5P_3P
-
RNA_HOTSPOT
-
GENE_EXPRESSION
-
RNAExonVariant
-
ProcControl
-
FLT3ITD
Note: The FLT3ITD variant type is detected only in Oncomine™ Myeloid Assay analysis workflows.
Filter conditions include the ability to select all variant types or specific variant types in filtered analysis results.
The filter returns variants in analyses that match the selected specific variant types.