Variant Classification filter
This filter can be added to filter chains in the software to narrow results to the following user-set variant classifications:
-
Unclassified
-
Unknown
-
Benign
-
Suspected Benign
-
Likely Benign
-
Suspected Deleterious
-
Deleterious
-
Pathogenic
-
Likely Pathogenic
-
VUS
-
Uncertain Significance
-
Technical Artifact
Filter conditions include the ability to include all (Filter Option set to In), none (Filter Option set to Not In), or to select specific classifications in filtered analysis results.
The filter returns variants that are classified by software users.