Default Fusions View filter
This filter can be added to filter chains in the software to detect all of the fusion variants, expression control markers, and assay 5′/3′ markers that the predefined Default Fusions View filter chain detects.
Filter conditions include the ability to select specific annotations (Present or Absent), to include all Default Fusions View variants (In), or to exclude all Default Fusions View variants (Not In) in filter analysis results. In addition, unannotated variants can be included or excluded in filtered analysis results.
The filter detects fusion variants, expression control markers, and assays 5′/3′ markers, which can be included or excluded in filtered analysis results.