Location filter
This filter can be added to filter chains in the software to detect variants in specific positions, such as exonic.
Filter conditions include the ability to select all locations or specific locations in filtered analysis results. Locations include:
-
unknown
-
intergenic
-
intronic
-
exonic
-
utr_5
-
utr_3
-
splicesite_5
-
splicesite_3
-
upstream
-
downstream
-
exonic_nc
-
intronic_nc
-
ncRNA
-
nonCoding
The filter returns variants in samples that match the selected filter locations.