Deamination filter
This filter can be added to filter chains in the software to filter C:G>T:A SNVs that are detected at all allele frequencies. Filter conditions include the ability to exclude (Not In) or include (In) these variants in filtered analysis results.