Allele Read-Count filter
This filter can be added to filter chains in the software to set the minimum count for genotype alleles.
Filter conditions include the ability to set allele read-count ranges from 0 to 100,000. In addition, the range boundary values can be included or excluded, and unannotated variants can be included or excluded in filtered analysis results.
The filter removes variant candidates that do not have the required numbers of supporting reads from analysis results.