Fusion Read Counts filter
This filter can be added to filter chains in the software to produce counts for fusion reads.
Filter conditions include the ability to set a minimum count of 0 or higher, to include the boundary value for the count, and to include or exclude unannotated variants in filtered analysis results.
This filter returns results that meet the fusion read minimum count value.