Genomic Coordinates filter
This filter can be added to filter chains in the software to narrow analysis results to specific genomic regions. The filter can also be used to identify regions that are targeted by an amplicon or amplicons in a user-defined assay.
The filter value is one chromosome region, or multiple regions that are separated by OR operators or commas.
The Genomic Coordinates filter returns variants in samples that are in the range for the genomic coordinates that you set.
IMPORTANT! Do not use AND, OR, or NOT when adding values to the Genomic Coordinates filter.