Oncomine filter
This filter is the default filter for most Oncomine assays. It presents the user with Oncomine driver variants that are based on their Oncomine Gene Class and Oncomine Variant Class annotations. It can be added to filter chains in the software to detect Oncomine-annotated variants in specific Oncomine panels.
Analysis workflows for Oncomine assays and analysis workflow templates for Ion AmpliSeq HD panels in Ion Reporter Software include the Oncomine Variant Annotator plugin. The plugin integrates into analysis results data from more than 24,000 exomes across solid tumor and hematological cancer types, and annotates variants relevant to cancer with Oncomine Gene class and Oncomine Variant class information. For more information and a full list of annotation rules for each Oncomine assay, see Oncomine Variant Annotator plugin criteria, or contact your local support representative, Field Bioinformatics Specialist (FBS), or Clinical Account Consultant (CAC).
The Oncomine filter (Oncomine = In) returns variants in the samples that have Oncomine Gene class and Oncomine Variant class annotation.
This filter can be used only if an Oncomine analysis workflow was used for the analysis. You can apply filter chains that include the Oncomine filter only if the analysis workflow that was used for the analysis includes the Oncomine Variant Annotator plugin. Examples of filter chains that include this filter are the Oncomine Variants and Oncomine Extended filter chains.