Variant Subtype filter
This filter can be added to filter chains in the software to detect variant subtypes, including:
-
BigDel
-
BigDup
-
GeneCNV
-
REF
-
NOCALL
Filter conditions include the ability to select all or specific variant subtypes in filtered analysis results.
The filter returns variants that match the selected specific variant subtypes.