Allele Ratio filter
This filter can be added to filter chains in the software to narrow analysis results to nonreference allele frequencies.
Filter conditions include the ability to set allele ratio ranges from 0.0 to 1.0. In addition, the range boundary values can be included or excluded, and unannotated variants can be included or excluded in filtered analysis results.
The filter returns all variants that have at least one nonreference allele reported with a frequency in the selected filter range.