Disease Research Area (DRA) filter
This filter can be added to filter chains in the software to annotate disease research categories as defined by the Disease Research Area (DRA) annotation source database. The specific filters are:
-
Disease Research Area (DRA)(20170112)
-
Disease Research Area (DRA)(20170914)
Filter conditions include the ability to select specific annotations, to include all Disease Research Area values (In), or to exclude all Disease Research Area values (Not In) in filtered analysis results. In addition, unannotated variants can be included or excluded in filtered analysis results.
This filter annotates disease research areas that match the DRA database, and can be included or excluded.