Remove filters
You can remove filters from the Analyses table in Ion Reporter Software.
  • Click  (Remove) next to the filter list to remove the filter from the Analyses table.
  • Click More Filters, then click the filter to remove the filter from the Analyses table.
  • Click More Filters, then click Clear to remove all of the selected filter categories from the list.
  • Click Clear All to remove all filters, then show all samples in the Analyses table.