Visualize an amplicon coverage report
To view an amplicon visually, click an amplicon link in the Amplicon Coverage Report screen to view the results in the Ion Reporter™ Genomic Viewer (IRGV) or the Integrative Genomics Viewer (IGV), depending on the viewer settings. For more information, see Set IRGV or IGV as the default viewer.
Figure 1. Amplicon visualization in IRGV