FusionSampleOverallCall
The FusionSampleOverallCall allows the user to confirm whether a sample is a No Call, Positive, or Negative. The following logic is applied for the panels that have both targeted fusion isoforms and 5'/3' Imbalance Assays:
|
Fusion Isoforms |
RET/ROS1/ALK Imbalance Score |
Final Call |
|---|---|---|
|
Positive |
Positive |
Positive |
|
Positive |
No Call/Negative |
Positive |
|
Negative |
Positive |
Positive |
|
Negative |
Positive |
Positive |
|
Negative |
No Call |
No Call |
|
Negative |
Negative |
Negative |