Jump to main content
Ion Reporter™ Software 5.22 Help
Logo
  1. Home
  2. Fusion analyses
  3. Confirm the presence of fusion calls in Ion Reporter™ Software
  • Introduction to Ion Reporter™ Software
  • Get started with Ion Reporter™ Software
  • Import and manage samples
  • Manage analysis workflows
  • Analyses
  • Analysis results
  • Detailed analysis metrics
  • Visualize analysis results with Ion Reporter™ Software
  • Create and view reports
  • Filters and filter chains
  • Annotation sources
  • Administer Ion Reporter™ Software
  • Troubleshooting
  • Fusion analyses
    • Fusion calls in analysis results
    • Analysis configuration file for gene fusion analysis
    • Data types for gene fusions analyses
    • Fusion sample QC metrics
    • Fusion detection methods
    • Confirm the presence of fusion calls in Ion Reporter™ Software
    • Filter chains for fusion panels
  • Ion Reporter™ Software web services API
  • Programming guidelines for Ion Reporter™ Software plugins
  • Oncomine™ Variant Annotator plugin criteria
  • Documentation and support
  • Glossary
  • Legal
Confirm the presence of fusion calls in Ion Reporter™ Software
Analysis results in Ion Reporter™ Software provide several ways to confirm a fusion call, including Imbalance values, visualization, and Normalized Detection Fractions. For more information, see Novel fusion detection using expression imbalance, View RNA Exon Tile Fusion Imbalance plots, and 3'/5' Imbalance scores for assay calls.
For Research Use Only. Not for use in diagnostic procedures.
thermofisher.com/support  |  thermofisher.com/askaquestion