View RNA Exon Tile Fusion Imbalance plots
You can identify novel fusions with predicted breakpoints of select driver genes in Ion Reporter™ Software with RNA Exon Tile Fusion Imbalance analysis results and plots.
You can view RNA Exon Tile Fusion Imbalance analysis results and plots from a single analysis or from multiple analyses simultaneously. If multiple analyses are compared, analysis results for the selected analyses are displayed side by side. Because each gene in each sample is normalized to baseline and read coverage, you can compare multiple samples from different chips, tissues, and so on.
- In the Analyses tab, click Overview.
- Select one or multiple analyses, then click Visualize.
-
In the Analysis Visualization screen, click RNA Exon Tile Fusion Imbalance.
The graphs and plots that are visual representations of RNA fusion imbalance analysis results are shown.
-
To change the analysis or analyses that are displayed, select a single analysis or select All selected analyses from the dropdown list.
RNA Exon Tile Fusion Imbalance plots are shown for a single analysis or all selected analyses. For example plots, see the following sections:
-
Example RNA Exon Tile Fusion Imbalance plots for a single analysis.
-
Example RNA Exon Tile Fusion Imbalance read count plot for a single analysis.
-
Example sample coverage plot for a single analysis positive for a fusion imbalance in NTRK1.
-
Example RNA Exon Tile Fusion Imbalance call plot for multiple analyses.
-
Example Normalized Read Depth by Gene RNA Exon Tile Fusion Imbalance plot for multiple analyses.
-
- (Optional) Click Download All Plots to download a ZIP file that contains all of the plots for all of the analyses shown in the RNA Exon Tile Fusion Imbalance visualization. For more information about file downloads, see View notifications.