Coverage histogram
When you zoom in on the variant pileup view, you see the coverage histogram and reads from the .bam track. The histogram in gray shows read depth at that location and reads from the .bam track show read direction and location of variants.
-
Read depth
-
Read location and direction of variants