Evaluate analysis results
You can evaluate analysis results in Ion Reporter™ Software by comparing multiple analyses. For example, you might compare new analysis results to known accurate analysis results.
Visualization of a single trio or paired analysis is supported, because trio and paired analyses each contain more than one set of variants.
  1. In the Analyses tab, click Overview.
  2. Select multiple analyses, then click Actions Visualize.
You can review the results in various ways. For more information, see Compare results of single or multiple analyses.