CNV Heat Map tab
The CNV Heat Map tab provides a visual representation of copy number variation (CNV) data in the form of a heat map. You can select multiple analyses to compare the CNV data for multiple samples side by side. Ploidy values are used to quantify and compare call number variations. The ploidy value of 2 is expected for wild type samples where no genetic amplification or deletion was detected. A ploidy value of >2 indicates amplification. A ploidy value of <2 indicates deletion. In the heat map, a color scale is used to visualize ploidy, where the expected value of 2 is colored gray. For X and Y chromosome, the color that is applied to the plot considers the expected value given the gender. For example, for a male, the expected value for Y is 1, therefore, the cell for that variant appears as gray.
Use the menus in the Filter Options pane to limit the data that is displayed in the heat map. For a large data set, the data for the first 2,000 genes are retrieved.
-
For more information about the Samples Venn diagram, see Samples Venn diagram in Filter Options.
-
For more information about using the Chromosome filter, see Chromosome filter in the Filter Options.
-
For more information about using the Filter Chains filter, see Filter Chains filter in the Filter Options.
-
Click Selected Analyses to view the list of the analyses that are visualized.
-
Heat map Filter Options.
-
Heat map legend: ploidy. Place the pointer over the legend the view the relationship between the ploidy value and the colors used in the heat map.
-
Samples and analyses: place the pointer over sample and analysis names to view the sample and analysis details.
-
Genes: place the pointer over a gene ID to view the details for that gene or click the gene ID to view the details for all analyses in IRGV. Gene order is determined by the chromosomal position of the gene. The chromosome number is displayed to the left of the gene list.
-
Variants: place the pointer over a variant to view the details pane for that variant, or click the cell to view the details for that variant in IRGV. If >1 CNVs are detected for the same gene, the number of CNVs is listed in the cell.
Use the following Chart Options to show the heat maps of choice and customize the heat map view.
|
Chart Option |
Description |
|---|---|
|
Scoring Function |
The CNV heat map uses ploidy values to score copy number variants. |
|
Gene Order |
The CNV heat map orders the list of genes based on their position on the chromosome. |
|
Sample Order |
Select one of following options from the Sample Order dropdown list to arrange the order in which the genes are listed in the heat map.
|
|
Color Scale |
Select a color scale to customize the colors that are displayed in the heat map. |
|
Export Heatmap |
You can export and save the heat map in PNG, SVG, and CSV file formats. The file is downloaded after you make a choice from the Export Heatmap dropdown list. |