Variants Table tab
The Variants Table lists the variant information for the analyses with Ion AmpliSeq™ workflows. If you selected more than one analysis, the Variants Table displays a side by side comparison of the variant calls for each analysis.
Use the menus in the Filter Options pane to limit the data that is displayed in the Variants Table.